PHOSPHORUS LIQUIRAPID 1 * 200 mL
Catalog # MSPP10027
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
EOSIN RAL 555 1L 1 * 1 L
Supplier: RAL
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
VWR® Rota® Duo, pH indicator papers
Supplier: VWR Chemicals
Test strips, peracetic acid, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Copper test strips, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Powder pillows, ammonia
Supplier: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach