Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
8AL Justiergaszylinder 15 ppm H2S / Rest N2 Aluminiumzylinder, 58 Liter nicht wiederbefüllbar 1 * 1.700 mL
Catalog # CALGC011610
Supplier:
CALGAZ
Expand image
8AL Justiergaszylinder 15 ppm H2S / Rest N2 Aluminiumzylinder, 58 Liter nicht wiederbefüllbar 1 * 1.700 mL
Catalog # CALGC011610
Supplier:
CALGAZ
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals