Biotech DEGASi PLUS Classic, 3-channels 480 µl Systec AF 1 * 1 items
Catalog # HPLC0003-6353-A
Supplier: COLLECTION PREFIX HPLC
Biotech DEGASi PLUS Classic, 3-channels 480 µl Systec AF 1 * 1 items
Catalog # HPLC0003-6353-A
Supplier: COLLECTION PREFIX HPLC
Frequently Bought Together
Short-term tubes for accuro®
Supplier: Dräger
Gas detection tube for hydrogen sulphide
Supplier: Dräger
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Electrode stand with swivel arm, Eutech
Supplier: EUTECH
VWR®, cell counting slides for automated cell counters
Supplier: VWR Collection
VWR®, Square Cuvettes
Supplier: VWR Collection
Gas detection tubes for water vapour
Supplier: Dräger
VWR®, Refractometers, Digital Handheld
Supplier: VWR Collection
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Accessories for titrators
Supplier: Mettler - Toledo
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
FlashGel™ DNA system
Supplier: LONZA
Accessories for CytoFLEX Platform
Supplier: Beckman Coulter
Plastic cuvette for LICO
Supplier: Hach