1-[2- (trifluoromethyl)pyrimidin5-yl]ethanone 1 * 5 g
Catalog # PHBLPBU2714
Supplier: Pharmablock (USA)
1-[2- (trifluoromethyl)pyrimidin5-yl]ethanone 1 * 5 g
Catalog # PHBLPBU2714
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals