TERT-BUTYL (4S)-3,3-DIFLUORO-4-HYDROXYPYRROLIDINE-1-CARBOXYLATE 1 * 2 g
Catalog # PHBLPBZ3796-2G
Supplier: Pharmablock (USA)
TERT-BUTYL (4S)-3,3-DIFLUORO-4-HYDROXYPYRROLIDINE-1-CARBOXYLATE 1 * 2 g
Catalog # PHBLPBZ3796-2G
Supplier: Pharmablock (USA)
Frequently Bought Together
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethylene glycol, Sigma-Aldrich®
Supplier: Merck
Formamide 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific