Cadence 316 SS fittings; male luer lock x 10-32 UNF thread 41507-31
MFLX41507-31
Exceptional durability.
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Constructed of durable 316 stainless steel, these Luer Adapter Fittings are ideal for a wide range of applications.
- Connection Type:Male Luer to Thread
- Flow pattern:Straight
- Description:316 SS fittings; male luer lock×10-32 UNF thread 41507-31
- Environmentally Preferable:
- Connection type (port 1):Male luer lock
- No. of ports:2
- Pk:1
- Fitting material:Stainless Steel 316
- Thread size UNF(M) (port 1):10-32
Frequently Bought Together