tert-butyl N-[(3R,4S)-3-fluoropiperidin-4-yl]carbamate - 97% 1 * 10 g
Catalog # PHBLPBZS1007-10G
Supplier: Pharmablock (USA)
tert-butyl N-[(3R,4S)-3-fluoropiperidin-4-yl]carbamate - 97% 1 * 10 g
Catalog # PHBLPBZS1007-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Butyl acetate 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Trichloroethylene 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.85% for HPLC
Supplier: Thermo Fisher Scientific
Trifluoroacetic Acid for UV 1 * 100 mL
Supplier: PanReac AppliChem