Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro spatula, straight, -, 150 mm, 4 mm
  RSGA80.531.004
 :   RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
  RSGA80.531.004
 :  RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro spatula, straight, -, 150 mm, 4 mm

 
20,70 €
   
1 * 1 items
 

Made from stainless steel 1.4301.

  • Rust-proof
  • Highly resistant against chemicals
  • With 6 edged grip for better handling
  • Autoclavable
 
  • Sterile:
    -
  • Description:
    Micro spatula, straight
  • Length:
    150 mm
  • Seq:
    05
  • Blade width:
    4 mm