Order Entry
Slovenia
ContactUsLinkComponent
Nickel nitrate matrix modifier solution 10 g/l Ni(NO3)2 (99,999 %) in 1 % HNO3 1 * 50 mL
Catalog # ROTH5146.1
Supplier:  Roth Carl
Nickel nitrate matrix modifier solution 10 g/l Ni(NO3)2 (99,999 %) in 1 % HNO3 1 * 50 mL
Catalog # ROTH5146.1
Supplier:  Roth Carl
Frequently Bought Together