Order Entry
Slovenia
ContactUsLinkComponent
Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (NUN-S47) was produced from a hybridoma resulting from the fusion of a mouse myelo ma with B cells obtained from a mouse immunized with purified recombinant SARS-CoV-2 Nucleocapsid protein. The IgG fr 1 * 2 mg
Catalog # ACRBNUN-S47-1.5MG
Supplier:  ACRO Biosystems
Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (NUN-S47) was produced from a hybridoma resulting from the fusion of a mouse myelo ma with B cells obtained from a mouse immunized with purified recombinant SARS-CoV-2 Nucleocapsid protein. The IgG fr 1 * 2 mg
Catalog # ACRBNUN-S47-1.5MG
Supplier:  ACRO Biosystems
Frequently Bought Together