Order Entry
Slovenia
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (NUN-C5145) is expressed from E.coli cells. It contains AA Ser 255 - Pro 364 (Accession # QHO62115.1). 1 * 1 mg
Catalog # ACRBNUN-C5145-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (NUN-C5145) is expressed from E.coli cells. It contains AA Ser 255 - Pro 364 (Accession # QHO62115.1). 1 * 1 mg
Catalog # ACRBNUN-C5145-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together