Order Entry
Slovenia
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein (R203M, D377Y), His Tag (NUN-C52Hn) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (R203M, D377Y)). The mutations (R203M, D377Y) on nucleocapsid protein were identif 1 * 100 µG
Catalog # ACRBNUN-C52HN100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein (R203M, D377Y), His Tag (NUN-C52Hn) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1 (R203M, D377Y)). The mutations (R203M, D377Y) on nucleocapsid protein were identif 1 * 100 µG
Catalog # ACRBNUN-C52HN100UG
Supplier:  ACRO Biosystems
Frequently Bought Together