Order Entry
Slovenia
ContactUsLinkComponent
Rat PSMA, His Tag (PSA-R5245) is expressed from human 293 cells (HEK293). It contains AA Lys 45 - Asp 752 (Accession # P70627-1). 1 * 100 µG
Catalog # ACRBPSA-R5245100UG
Supplier:  ACRO Biosystems
Rat PSMA, His Tag (PSA-R5245) is expressed from human 293 cells (HEK293). It contains AA Lys 45 - Asp 752 (Accession # P70627-1). 1 * 100 µG
Catalog # ACRBPSA-R5245100UG
Supplier:  ACRO Biosystems
Frequently Bought Together