Order Entry
Slovenia
ContactUsLinkComponent
Human FGL1 (64-312), Fc Tag (FG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08 830-1). 1 * 100 µG
Catalog # ACRBFG1-H5254100UG
Supplier:  ACRO Biosystems
Human FGL1 (64-312), Fc Tag (FG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08 830-1). 1 * 100 µG
Catalog # ACRBFG1-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together