Order Entry
Slovenia
ContactUsLinkComponent
Cynomolgus APRIL, His,Flag Tag (APL-C52D2) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # XP_005582818.1). 1 * 1 mg
Catalog # ACRBAPL-C52D2-1MG
Supplier:  ACRO Biosystems
Cynomolgus APRIL, His,Flag Tag (APL-C52D2) is expressed from human 293 cells (HEK293). It contains AA Gln 111 - Leu 250 (Accession # XP_005582818.1). 1 * 1 mg
Catalog # ACRBAPL-C52D2-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together