Cynomolgus / Rhesus macaque Cadherin-6, His Tag (CA6-C52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Gly 614 (Accession # G7MUQ7-1). 1 * 1 mg
Catalog # ACRBCA6-C52H3-1MG
Supplier: ACRO Biosystems
Frequently Bought Together