Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Cynomolgus LILRB2, His,Avitag (LI2-C82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 ( Accession # BAE87348.1). 1 * 25 µG
Catalog # ACRBLI2-C82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus LILRB2, His,Avitag (LI2-C82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 ( Accession # BAE87348.1). 1 * 25 µG
Catalog # ACRBLI2-C82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together