Order Entry
Slovenia
ContactUsLinkComponent
Cynomolgus CD79B, His Tag (CDB-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 161 (Accession # A0A2K 5WTH8-1). 1 * 1 mg
Catalog # ACRBCDB-C52H3-1MG
Supplier:  ACRO Biosystems
Cynomolgus CD79B, His Tag (CDB-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 30 - Asp 161 (Accession # A0A2K 5WTH8-1). 1 * 1 mg
Catalog # ACRBCDB-C52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together