Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

4-Hydroxyindole

Bottle, 25 g
Catalog # APOSOR10198-25G
Irritant
Supplier:   Apollo Scientific
4-Hydroxyindole
Catalog # APOSOR10198-25G
Supplier:  Apollo Scientific
CAS Number:  2380-94-1
4-Hydroxyindole

Bottle, 25 g

CAS Number:  2380-94-1
Price
145,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
  • CAS Number:2380-94-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00005667
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:150 °C/4mmHg
  • Melting Pt:97…99 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Cat. No.:
    APOSOR10198-25G

Specifications