Order Entry
Slovenia
Orders LinkContactUsLinkComponent
4-Hydroxyindole

4-Hydroxyindole

Bottle, 1 g
Catalog # APOSOR10198-1G
Irritant
Supplier:   Apollo Scientific
4-Hydroxyindole
Catalog # APOSOR10198-1G
Supplier:  Apollo Scientific
CAS Number:  2380-94-1
4-Hydroxyindole

Bottle, 1 g

CAS Number:  2380-94-1
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
  • CAS Number:2380-94-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00005667
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:150 °C/4mmHg
  • Melting Pt:97…99 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.