Order Entry
Slovenia
ContactUsLinkComponent
Human PD-L1, His Tag PD1-H5229 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Arg 238 Accession NP_054862.1 . 1 * 100 µG
Catalog # ACRBPD1-H5229100UG
Supplier:  ACRO Biosystems
Human PD-L1, His Tag PD1-H5229 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Arg 238 Accession NP_054862.1 . 1 * 100 µG
Catalog # ACRBPD1-H5229100UG
Supplier:  ACRO Biosystems
Frequently Bought Together