Order Entry
Slovenia
ContactUsLinkComponent
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.9 kDa. The protein migrates as 60- 80 kDa under reducing R condition SDS-PAGE due to glycosylation. 1 * 96 Tests
Catalog # ACRBEBS-001-96TEST
Supplier:  ACRO Biosystems
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.9 kDa. The protein migrates as 60- 80 kDa under reducing R condition SDS-PAGE due to glycosylation. 1 * 96 Tests
Catalog # ACRBEBS-001-96TEST
Supplier:  ACRO Biosystems
Frequently Bought Together