Order Entry
Slovenia
ContactUsLinkComponent
The biotinylated EGFR protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-EGFR antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 2 mg
Catalog # ACRBMBE-K012-2MG
Supplier:  ACROBIOSYSTEMS
The biotinylated EGFR protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-EGFR antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 2 mg
Catalog # ACRBMBE-K012-2MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together