Order Entry
Slovenia
ContactUsLinkComponent
Cynomolgus IL-23A Mouse IL-12B Heterodimer Protein, His TagTag Free ILB-CM52W8 is expressed from human 293 cells HEK293. It contain s AA Val 22 - Pro 189 IL23A Met 23 - Ser 335 IL12B Accession A0A2K5TLV4-1 IL23A P43432-1 IL12B. 1 * 50 µG
Catalog # ACRBILB-CM52W850UG
Supplier:  ACRO Biosystems
Cynomolgus IL-23A Mouse IL-12B Heterodimer Protein, His TagTag Free ILB-CM52W8 is expressed from human 293 cells HEK293. It contain s AA Val 22 - Pro 189 IL23A Met 23 - Ser 335 IL12B Accession A0A2K5TLV4-1 IL23A P43432-1 IL12B. 1 * 50 µG
Catalog # ACRBILB-CM52W850UG
Supplier:  ACRO Biosystems
Frequently Bought Together