Order Entry
Slovenia
ContactUsLinkComponent
Mouse NOTCH1, His Tag NO1-M52H8 is expressed from human 293 cells HEK293. It contains AA Ala 19 - Gln 526 Accession Q01705-1. 1 * 1 mg
Catalog # ACRBNO1-M52H8-1MG
Supplier:  ACRO Biosystems
Mouse NOTCH1, His Tag NO1-M52H8 is expressed from human 293 cells HEK293. It contains AA Ala 19 - Gln 526 Accession Q01705-1. 1 * 1 mg
Catalog # ACRBNO1-M52H8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together