Order Entry
Slovenia
ContactUsLinkComponent
FITC-Labeled Human CD5, His Tag CD5-H52H2 is expressed from human 293 cells HEK293. It contains AA Arg 25 - Pro 372 Accession P0612 7-1. It is the FITC labeled form of Human CD5, His Tag CD5-H52H5. 1 * 200 µG
Catalog # ACRBCD5-H52H2200UG
Supplier:  ACRO Biosystems
FITC-Labeled Human CD5, His Tag CD5-H52H2 is expressed from human 293 cells HEK293. It contains AA Arg 25 - Pro 372 Accession P0612 7-1. It is the FITC labeled form of Human CD5, His Tag CD5-H52H5. 1 * 200 µG
Catalog # ACRBCD5-H52H2200UG
Supplier:  ACRO Biosystems
Frequently Bought Together