Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG2a Fc,Avitag SPD-C82A9 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accession QHD43416.1. 1 * 25 µG
Catalog # ACRBSPD-C82A9-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG2a Fc,Avitag SPD-C82A9 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accession QHD43416.1. 1 * 25 µG
Catalog # ACRBSPD-C82A9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together