Order Entry
Slovenia
ContactUsLinkComponent
SARS-CoV-2 S protein RBD N440K, His Tag SRD-C52H2 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 N440K. 1 * 1 mg
Catalog # ACRBSRD-C52H2-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD N440K, His Tag SRD-C52H2 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 N440K. 1 * 1 mg
Catalog # ACRBSRD-C52H2-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together