Order Entry
Slovenia
ContactUsLinkComponent
FITC-Labeled Human Nectin-4, His Tag NE4-HF2H3 is expressed from human 293 cells HEK293. It contains AA Gly 32 - Ser 349 Accession Q96NY8-1. It is the FITC labeled form of Human Nectin-4, His Tag NE4-H52H3. 1 * 200 µG
Catalog # ACRBNE4-HF2H3200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human Nectin-4, His Tag NE4-HF2H3 is expressed from human 293 cells HEK293. It contains AA Gly 32 - Ser 349 Accession Q96NY8-1. It is the FITC labeled form of Human Nectin-4, His Tag NE4-H52H3. 1 * 200 µG
Catalog # ACRBNE4-HF2H3200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together