Order Entry
Slovenia
ContactUsLinkComponent
Human ZNRF3, His Tag ZN3-H52H3 is expressed from human 293 cells HEK293. It contains AA Lys 56 - Met 219 Accession Q9ULT6-1. 1 * 100 µG
Catalog # ACRBZN3-H52H3100UG
Supplier:  ACRO Biosystems
Human ZNRF3, His Tag ZN3-H52H3 is expressed from human 293 cells HEK293. It contains AA Lys 56 - Met 219 Accession Q9ULT6-1. 1 * 100 µG
Catalog # ACRBZN3-H52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together