Order Entry
Slovenia
ContactUsLinkComponent
Human Tissue Factor, Fc Tag TF3-H5253 is expressed from human 293 cells HEK293. It contains AA Ser 33 - Glu 251 Accession P13726-1. 1 * 100 µG
Catalog # ACRBTF3-H5253100UG
Supplier:  ACRO Biosystems
Human Tissue Factor, Fc Tag TF3-H5253 is expressed from human 293 cells HEK293. It contains AA Ser 33 - Glu 251 Accession P13726-1. 1 * 100 µG
Catalog # ACRBTF3-H5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together