Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human NKG2CCD94, His,Avitag NC4-H82E3 is expressed from human 293 cells HEK293. It contains AA Ile 94 - Leu 231 NKG2C Asp 57 - Ile 179 CD94 Accession P26717-1 NKG2C Q13241-1 CD94. 1 * 200 µG
Catalog # ACRBNC4-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human NKG2CCD94, His,Avitag NC4-H82E3 is expressed from human 293 cells HEK293. It contains AA Ile 94 - Leu 231 NKG2C Asp 57 - Ile 179 CD94 Accession P26717-1 NKG2C Q13241-1 CD94. 1 * 200 µG
Catalog # ACRBNC4-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together