Order Entry
Slovenia
ContactUsLinkComponent
Human PD-L1 19-134, His Tag PD1-H52H3 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Tyr 134 Accession Q9NZQ7-1. 1 * 100 µG
Catalog # ACRBPD1-H52H3100UG
Supplier:  ACRO Biosystems
Human PD-L1 19-134, His Tag PD1-H52H3 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Tyr 134 Accession Q9NZQ7-1. 1 * 100 µG
Catalog # ACRBPD1-H52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together