Order Entry
Slovenia
ContactUsLinkComponent
Canine CD22, His Tag CD2-C52H3 is expressed from human 293 cells HEK293. It contains AA Met 1 - Arg 718 Accession F1PN92-1. 1 * 1 mg
Catalog # ACRBCD2-C52H3-1MG
Supplier:  ACRO Biosystems
Canine CD22, His Tag CD2-C52H3 is expressed from human 293 cells HEK293. It contains AA Met 1 - Arg 718 Accession F1PN92-1. 1 * 1 mg
Catalog # ACRBCD2-C52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together