Order Entry
Slovenia
ContactUsLinkComponent
Human latent GDF-8, His Tag GD8-H5243 is expressed from human 293 cells HEK293. It contains AA Asn 24 - Ser 375 Accession O14793-1. 1 * 500 µG
Catalog # ACRBGD8-H5243500UG
Supplier:  ACRO Biosystems
Human latent GDF-8, His Tag GD8-H5243 is expressed from human 293 cells HEK293. It contains AA Asn 24 - Ser 375 Accession O14793-1. 1 * 500 µG
Catalog # ACRBGD8-H5243500UG
Supplier:  ACRO Biosystems
Frequently Bought Together