Order Entry
Slovenia
ContactUsLinkComponent
Rabbit CD3ECD3D Heterodimer Protein, His TagFlag Tag CDD-R52D7 is expressed from human 293 cells HEK293. It contains AA Gln 22 - Thr 120 CD3E Thr 22 - Ser 107 CD3D Accession Q9TUF9-1 CD3E A0A5F9D3I4-1 CD3D. 1 * 1 mg
Catalog # ACRBCDD-R52D7-1MG
Supplier:  ACRO Biosystems
Rabbit CD3ECD3D Heterodimer Protein, His TagFlag Tag CDD-R52D7 is expressed from human 293 cells HEK293. It contains AA Gln 22 - Thr 120 CD3E Thr 22 - Ser 107 CD3D Accession Q9TUF9-1 CD3E A0A5F9D3I4-1 CD3D. 1 * 1 mg
Catalog # ACRBCDD-R52D7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together