Order Entry
Slovenia
ContactUsLinkComponent
SARS-CoV-2 S1 protein NTD, His Tag S1D-C52Hc is expressed from human 293 cells HEK293. It contains AA Ser 13 - Leu 303 Accession QH D43416.1 L18F, D80A, D215G, 242-244del, R246I. 1 * 100 µG
Catalog # ACRBS1D-C52HC100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein NTD, His Tag S1D-C52Hc is expressed from human 293 cells HEK293. It contains AA Ser 13 - Leu 303 Accession QH D43416.1 L18F, D80A, D215G, 242-244del, R246I. 1 * 100 µG
Catalog # ACRBS1D-C52HC100UG
Supplier:  ACRO Biosystems
Frequently Bought Together