Order Entry
Slovenia
ContactUsLinkComponent
THE AUTO SIPPER ACCESSORY (N4101012) CONSISTS OF AN AUTOMATIC PERISTALTIC PUMP, A PUMPING CARTRIDGE, TUBING, AND A COMPUTER INTERFAC E CABLE. THIS SIPPER SYSTEM USES SOFTWARE TO CONTROL THE AUTOMATION AND FLOW OF SAMPLE LIQUIDS. IT HAS TWO FLOW CHANN 1 * 1 items
Catalog # PERCN4101051
Supplier:  PerkinElmer
THE AUTO SIPPER ACCESSORY (N4101012) CONSISTS OF AN AUTOMATIC PERISTALTIC PUMP, A PUMPING CARTRIDGE, TUBING, AND A COMPUTER INTERFAC E CABLE. THIS SIPPER SYSTEM USES SOFTWARE TO CONTROL THE AUTOMATION AND FLOW OF SAMPLE LIQUIDS. IT HAS TWO FLOW CHANN 1 * 1 items
Catalog # PERCN4101051
Supplier:  PerkinElmer
Frequently Bought Together