Order Entry
Slovenia
ContactUsLinkComponent
Stainless steel slide-in drip tray, 15 mm rim, with guide bars and fixing screws (may affect the temperature distribution) - can be used only in connection with option K1 1 * 1 items
Catalog # MEMMB32763UF450
Supplier:  MEMMERT
Stainless steel slide-in drip tray, 15 mm rim, with guide bars and fixing screws (may affect the temperature distribution) - can be used only in connection with option K1 1 * 1 items
Catalog # MEMMB32763UF450
Supplier:  MEMMERT
Frequently Bought Together