Order Entry
Slovenia
ContactUsLinkComponent
SpectraMax AlphaScreen 384 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen assays. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, a patent pending design e a 1 * 1 items
Catalog # MLDV0200-7018
Supplier:  Molecular Devices
SpectraMax AlphaScreen 384 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScreen assays. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. In addition, a patent pending design e a 1 * 1 items
Catalog # MLDV0200-7018
Supplier:  Molecular Devices
Frequently Bought Together