Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human IGF-II, Avitag,Fc Tag (IG2-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Acc ession P01344-1). 1 * 200 µG
Catalog # ACRBIG2-H82F9200UG
Supplier:  ACRO Biosystems
Biotinylated Human IGF-II, Avitag,Fc Tag (IG2-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Acc ession P01344-1). 1 * 200 µG
Catalog # ACRBIG2-H82F9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together