Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human PD-L1, Avitag,His Tag (recommended for biopanning) (PD1-H82E5) is expressed from human 293 cells (HEK293). It con tains AA Phe 19 - Arg 238 (Accession NP054862.1). 1 * 25 µG
Catalog # ACRBPD1-H82E5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human PD-L1, Avitag,His Tag (recommended for biopanning) (PD1-H82E5) is expressed from human 293 cells (HEK293). It con tains AA Phe 19 - Arg 238 (Accession NP054862.1). 1 * 25 µG
Catalog # ACRBPD1-H82E5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together