Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human EGF R, His,Avitag (EGR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Access ion P00533-1). 1 * 25 µG
Catalog # ACRBEGR-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human EGF R, His,Avitag (EGR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Access ion P00533-1). 1 * 25 µG
Catalog # ACRBEGR-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together