Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human TRAIL R2, Avitag,His Tag (TR2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession O14763-1). 1 * 200 µG
Catalog # ACRBTR2-H82E6200UG
Supplier:  ACRO Biosystems
Biotinylated Human TRAIL R2, Avitag,His Tag (TR2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Ile 56 - Glu 182 (Accession O14763-1). 1 * 200 µG
Catalog # ACRBTR2-H82E6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together