Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human SIRP alpha, Fc,Avitag (CDA-H82F2) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Ac cession P78324-1). 1 * 200 µG
Catalog # ACRBCDA-H82F2200UG
Supplier:  ACRO Biosystems
Biotinylated Human SIRP alpha, Fc,Avitag (CDA-H82F2) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Ac cession P78324-1). 1 * 200 µG
Catalog # ACRBCDA-H82F2200UG
Supplier:  ACRO Biosystems
Frequently Bought Together