Mouse recombinant BAFF (from HEK293)
100 µG
Catalog # ACRBBAF-M5257100UG
Supplier: ACRO Biosystems
Mouse recombinant BAFF (from HEK293)
Catalog # ACRBBAF-M5257100UG
Supplier: ACRO Biosystems
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:100 µG
- Protease-free:Y
- Protein/peptide name:BAFF
- Protein synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|BLYS|BAFF
- Purity:>95%
- Endotoxin level:Low
- Source:HEK293 cells
- Biological activity:Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Mouse BAFF, Mouse IgG2a Fc Tag at 5 μg/ml (100 µl/well) can bind Mouse BCMA, Fc Tag with a linear range of 2-10 ng/ml (QC tested).
- Molecular weight:47,7 kDa
- Shipping temperature:Powder,RT
- Nuclease-free:Y
- Cat. No.:ACRBBAF-M5257100UG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Storage conditions:–20 °C
- Gene ID:Q9WU72-1
- Carrier-free:Y
- Formulation:Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Conjugation:Unconjugated
- Species:Mouse
- Animal-free:Y



