Mouse recombinant BAFF (from HEK293)
1 mg
Catalog # ACRBBAF-M5257-1MG
Supplier: ACRO Biosystems
Mouse recombinant BAFF (from HEK293)
Catalog # ACRBBAF-M5257-1MG
Supplier: ACRO Biosystems
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 mg
- Protease-free:Y
- Protein/peptide name:BAFF
- Protein synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|BLYS|BAFF
- Purity:>95%
- Endotoxin level:Low
- Source:HEK293 cells
- Biological activity:Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. Immobilised Mouse BAFF, Mouse IgG2a Fc Tag at 5 μg/ml (100 µl/well) can bind Mouse BCMA, Fc Tag with a linear range of 2-10 ng/ml (QC tested).
- Molecular weight:47,7 kDa
- Shipping temperature:Powder,RT
- Nuclease-free:Y
- Cat. No.:ACRBBAF-M5257-1MG
- Protein/peptide type:Recombinant
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Storage conditions:–20 °C
- Gene ID:Q9WU72-1
- Carrier-free:Y
- Formulation:Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Conjugation:Unconjugated
- Species:Mouse
- Animal-free:Y
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



