Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human SCF, Avitag,His Tag (SCF-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Ala 190 (Acce ssion AAH69797). 1 * 200 µG
Catalog # ACRBSCF-H82E1200UG
Supplier:  ACRO Biosystems
Biotinylated Human SCF, Avitag,His Tag (SCF-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 26 - Ala 190 (Acce ssion AAH69797). 1 * 200 µG
Catalog # ACRBSCF-H82E1200UG
Supplier:  ACRO Biosystems
Frequently Bought Together