Order Entry
Slovenia
ContactUsLinkComponent
Mouse IL-15, Fc Tag (IL5-M5255) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession P48346-1). 1 * 50 µG
  ACRBIL5-M5255-50UG
Mouse IL-15, Fc Tag (IL5-M5255) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession P48346-1). 1 * 50 µG
  ACRBIL5-M5255-50UG
Frequently Bought Together