Order Entry
Slovenia
ContactUsLinkComponent
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 200 µG
Catalog # ACRBPRN-H82F7200UG
Supplier:  ACRO Biosystems
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 200 µG
Catalog # ACRBPRN-H82F7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together